Assay Details
| Gene Symbol: Mrpl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172252.3 | NP_758456.2 | 5 - 6 | 489 | 94 | |
| GenBank mRNA | AK003285.1 | - | 5 - 6 | 476 | 94 | |
| AK012784.1 | - | 5 - 6 | 481 | 94 | ||
| AK020739.1 | - | 5 - 6 | 490 | 94 | ||
| BC028768.1 | - | 5 - 6 | 464 | 1282313 | 94 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrpl21
Genomic Map