Assay Details
| Gene Symbol: Iah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026347.3 | NP_080623.2 | 3 - 4 | 323 | 78 | |
| GenBank mRNA | AK005287.1 | - | 3 - 4 | 324 | 78 | |
| AK011299.1 | - | 3 - 4 | 324 | 78 | ||
| AK014743.1 | - | 3 - 4 | 301 | 78 | ||
| BC060949.1 | - | 3 - 4 | 326 | 6533986 | 78 | |
| BC087901.1 | - | 3 - 4 | 294 | 30303437 | 78 | |
More Information
Set Membership: |
|

Iah1
Genomic Map