Assay Details
| Gene Symbol: Dach1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038610.2 | NP_001033699.1 | 7 - 8 | 2155 | 69 | |
| NM_007826.3 | NP_031852.1 | 8 - 9 | 2311 | 69 | ||
| GenBank mRNA | AF129510.1 | - | 7 - 8 | 1917 | 69 | |
| AK047409.1 | - | 7 - 8 | 1021 | 69 | ||
| AK053594.1 | - | 7 - 8 | 1364 | 69 | ||
| AK139060.1 | - | 7 - 8 | 2156 | 69 | ||
| BC059934.1 | - | 7 - 8 | 1933 | 69 | ||
| BC078644.1 | - | 7 - 8 | 1689 | 69 | ||
| BC145440.1 | - | 7 - 8 | 1761 | 69 | ||
More Information
Set Membership: |
|

Dach1
Genomic Map