Assay Details
Gene Symbol: Cdk2ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026373.4 | NP_080649.1 | 3 - 4 | 392 | 75 | |
GenBank mRNA | AK003686.2 | - | 3 - 4 | 358 | 75 | |
AK012121.1 | - | 3 - 4 | 395 | 75 | ||
AK018035.1 | - | 3 - 4 | 392 | 75 | ||
BC025656.1 | - | 3 - 4 | 358 | 5342719 | 75 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921020 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921018 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921019 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921021 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921023 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921022 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921016 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921017 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P111259 | AK018035.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P111259 | AK181155.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P111259 | BC025656.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P111259 | NM_026373.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P111259 | AK003686.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P111259 | AK018035.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P111259 | AK181155.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P111259 | BC025656.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P111259 | NM_026373.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P111259 | AK003686.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 286256 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P111259 | AK018035.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P111259 | AK181155.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P111259 | BC025656.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P111259 | NM_026373.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P111259 | AK003686.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P111259 | AK018035.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P111259 | AK181155.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P111259 | BC025656.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P111259 | NM_026373.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P111259 | AK003686.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921028 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921036 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921033 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920996 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921032 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920998 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921006 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921027 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921022 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921030 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921024 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921023 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921013 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921008 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921016 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921020 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921011 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921001 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921015 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920994 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921005 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921003 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921026 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921007 | - | - |
Whole Mouse Genome G4122A | A_51_P111259 | AK018035.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P111259 | AK181155.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P111259 | BC025656.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P111259 | NM_026373.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P111259 | AK003686.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P111259 | AK018035.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P111259 | AK181155.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P111259 | BC025656.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P111259 | NM_026373.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P111259 | AK003686.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128415 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140547 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14179903 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14380315 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14380319 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14286452 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14286449 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14380345 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921018 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921020 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921023 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921022 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921021 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921016 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921019 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921042 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31920994 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921030 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921003 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14175733 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14179903 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921011 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921004 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31920996 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921023 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31920982 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31920985 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921024 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14390367 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921036 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14224496 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31920978 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921033 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31920988 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921007 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921020 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14349121 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921006 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921001 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31920990 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921032 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14380355 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921028 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31920995 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921014 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31920998 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31920983 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14402125 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14355203 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31920977 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921017 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10460316 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10460313 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10460315 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10460314 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417161_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1456490_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10460312 | - | - |
Mouse Genome 430 2.0 Array | 1456490_at | - | - |
Mouse Genome 430 2.0 Array | 1417161_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1456490_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1417161_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52004_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52004_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1900000331.mm.1 | NM_026373.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900000331.mm.1 | NM_026373.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900000331.mm.1 | TC1900000026.mm.1 | MTA TC |
Mouse Expression Set 430 | 1417161_PM_at | - | - |
Mouse Expression Set 430 | 1417161_at | - | - |
Mouse Expression Set 430 | 1456490_at | - | - |
Mouse Expression Set 430 | 1456490_PM_at | - | - |
Murine Genome U74v2 Set | 95679_at | - | - |
Murine Genome U74v2 Set | 95679_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1417161_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1417161_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417161_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417161_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456490_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456490_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 931627 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4070577 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004070577 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2627093 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2627093 | - | - |
mouse-6 v1.1 (Illumina) | scl52801.4.1_26-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4070577 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl52801.4.1_26-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4070577 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2627093 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2627093 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl52801.4.1_26-S | - | - |
