Assay Details
Gene Symbol: Cdk2ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026373.4 | NP_080649.1 | 1 - 2 | 159 | 133 | |
GenBank mRNA | AK003686.2 | - | 1 - 2 | 125 | 133 | |
AK012121.1 | - | 1 - 2 | 162 | 133 | ||
AK018035.1 | - | 1 - 2 | 159 | 133 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK003686.2, AK012121.1, AK018035.1, NM_026373.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
