Assay Details
| Gene Symbol: Ppp1ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031868.2 | NP_114074.1 | 1 - 2 | 98 | 77 | |
| GenBank mRNA | AK007932.1 | - | 1 - 2 | 99 | 77 | |
| AK028392.1 | - | 1 - 2 | 88 | 77 | ||
| AK090070.1 | - | 1 - 2 | 116 | 77 | ||
| AK150344.1 | - | 1 - 2 | 116 | 77 | ||
| AK151582.1 | - | 1 - 2 | 116 | 77 | ||
| AK152051.1 | - | 1 - 2 | 116 | 77 | ||
| AK152585.1 | - | 1 - 2 | 116 | 77 | ||
| AK152667.1 | - | 1 - 2 | 94 | 77 | ||
| AK153517.1 | - | 1 - 2 | 80 | 77 | ||
| AK159575.1 | - | 1 - 2 | 106 | 77 | ||
| AK167244.1 | - | 1 - 2 | 99 | 77 | ||
| AK167538.1 | - | 1 - 2 | 111 | 77 | ||
| AK167880.1 | - | 1 - 2 | 100 | 77 | ||
| AK167981.1 | - | 1 - 2 | 99 | 77 | ||
| BC014828.1 | - | 1 - 2 | 104 | 4239005 | 77 | |
| U25809.1 | - | 1 - 2 | 119 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ppp1ca
Genomic Map