Assay Details
| Gene Symbol: Cnih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB006191.2 | - | 5 - 6 | 616 | 101 | |
| AK141483.1 | - | 5 - 6 | 716 | 101 | ||
| BC059922.1 | - | 5 - 6 | 702 | 101 | ||
| BC103797.1 | - | 5 - 6 | 690 | 30134114 | 101 | |
| BC137585.1 | - | 5 - 6 | 577 | 8860604 | 101 | |
| BC137586.1 | - | 5 - 6 | 577 | 8860605 | 101 | |
| BC144739.1 | - | 5 - 6 | 603 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976033 | BC103797.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976033 | AK141483.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976033 | BC059922.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976033 | NM_009920.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976033 | AB006191.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2172956 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630234 | AK041529.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 849311 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 940627 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159767 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976033 | BC103797.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976033 | AK141483.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976033 | BC059922.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976033 | NM_009920.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976033 | AB006191.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2172956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926887 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159767 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03542389 | - | - |
| Whole Mouse Genome G4122A | A_51_P204801 | BC103797.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P204801 | AK141483.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P204801 | BC059922.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P204801 | NM_009920.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P204801 | AB006191.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125871 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130089 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159767 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14438353 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14478863 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14438360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14478854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14228236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14478843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926905 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926909 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14478848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14228242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14228220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14440973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926873 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14228217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14440970 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10464918 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464921 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464922 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464919 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464920 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464923 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10464917 | - | - |
| Mouse Genome 430 2.0 Array | 1417524_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417524_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12794_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12794_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900008385.mm.1 | NM_009920.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900008385.mm.1 | TC1900000951.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900008386.mm.1 | NM_009920.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900008386.mm.1 | TC1900000951.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417524_at | - | - |
| Mouse Expression Set 430 | 1417524_PM_at | - | - |
| Murine Genome U74v2 Set | 160830_PM_at | - | - |
| Murine Genome U74v2 Set | 160830_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417524_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417524_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5080609 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5080609 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005080609 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1252776 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1252776 | - | - |
| mouse-6 v1.1 (Illumina) | scl53484.6.1_117-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5080609 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53484.6.1_117-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5080609 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1252776 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1252776 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl53484.6.1_117-S | - | - |

Cnih2
Genomic Map