Assay Details
Gene Symbol: Drap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291080.1 | NP_001278009.1 | 1 - 2 | 311 | 76 | |
NM_024176.2 | NP_077138.1 | 1 - 2 | 311 | 76 | ||
GenBank mRNA | AK010152.1 | - | 1 - 2 | 265 | 76 | |
AK087456.1 | - | 1 - 2 | 137 | 76 | ||
AK154251.1 | - | 91 | 76 | |||
BC002090.1 | - | 1 - 2 | 248 | 3488139 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK010152.1, AK087456.1, AK154251.1, BC002090.1, NM_001291080.1, NM_024176.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
