Assay Details
| Gene Symbol: Drap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291080.1 | NP_001278009.1 | 5 - 6 | 711 | 87 | |
| NM_024176.2 | NP_077138.1 | 5 - 6 | 690 | 87 | ||
| GenBank mRNA | AK005445.1 | - | 4 - 5 | 425 | 87 | |
| AK010152.1 | - | 5 - 6 | 644 | 87 | ||
| AK087456.1 | - | 5 - 6 | 516 | 87 | ||
| AK154251.1 | - | 4 - 5 | 470 | 87 | ||
| BC002090.1 | - | 5 - 6 | 627 | 3488139 | 87 | |
| CT010290.1 | - | 5 - 6 | 425 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Drap1
Genomic Map