Assay Details
Gene Symbol: Capn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110504.1 | NP_001103974.1 | 8 - 9 | 1021 | 83 | |
NM_007600.3 | NP_031626.1 | 8 - 9 | 1073 | 83 | ||
GenBank mRNA | AF021847.1 | - | 8 - 9 | 952 | 83 | |
AF084459.1 | - | 7 - 8 | 939 | 83 | ||
AK088547.1 | - | 8 - 9 | 993 | 83 | ||
AK149090.1 | - | 4 - 5 | 421 | 83 | ||
AK149706.1 | - | 8 - 9 | 1013 | 83 | ||
AK150084.1 | - | 8 - 9 | 1012 | 83 | ||
AK160940.1 | - | 8 - 9 | 1013 | 83 | ||
AK165258.1 | - | 8 - 9 | 1032 | 83 | ||
AK168063.1 | - | 8 - 9 | 1012 | 83 | ||
AK171405.1 | - | 8 - 9 | 976 | 83 | ||
BC026138.1 | - | 8 - 9 | 988 | 4218148 | 83 | |
BC050276.1 | - | 8 - 9 | 989 | 4923347 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
