Assay Details
| Gene Symbol: Tgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029578.3 | NP_083854.3 | 8 - 9 | 696 | 66 | |
| GenBank mRNA | AK011439.1 | - | 7 - 8 | 584 | 66 | |
| AK011555.1 | - | 8 - 9 | 687 | 66 | ||
| AK154341.1 | - | 8 - 9 | 673 | 66 | ||
| AK165685.1 | - | 7 - 8 | 579 | 66 | ||
| AK165955.1 | - | 8 - 9 | 680 | 66 | ||
| BC021419.1 | - | 8 - 9 | 672 | 5012877 | 66 | |
More Information
Set Membership: |
|

Tgds
Genomic Map