Assay Details
| Gene Symbol: Acpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019807.2 | NP_062781.2 | 5 - 6 | 619 | 178 | |
| NM_207668.2 | NP_997551.1 | 5 - 6 | 619 | 178 | ||
| GenBank mRNA | AF210243.1 | - | 5 - 6 | 572 | 178 | |
| AK020693.1 | - | 5 - 6 | 580 | 179 | ||
| AK029273.1 | - | 5 - 6 | 605 | 178 | ||
| BC139826.1 | - | 5 - 6 | 610 | 40142642 | 178 | |
More Information
Set Membership: |
|

Acpp
Genomic Map