Assay Details
| Gene Symbol: Nudt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029385.2 | NP_083661.2 | 1 - 2 | 161 | 129 | |
| GenBank mRNA | AK009737.1 | - | 1 - 2 | 162 | 129 | |
| BC064048.1 | - | 1 - 2 | 148 | 574013 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nudt16
Genomic Map