| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900002184.mm.1 |
NM_001286103.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002184.mm.1 |
TC1900000163.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900002181.mm.1 |
NM_001286103.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002181.mm.1 |
TC1900000163.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900002182.mm.1 |
NM_001286103.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002182.mm.1 |
TC1900000163.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900002191.mm.1 |
NM_001286103.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002191.mm.1 |
NM_025463.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002191.mm.1 |
NR_104416.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002191.mm.1 |
TC1900000163.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900002183.mm.1 |
NM_001286103.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002183.mm.1 |
TC1900000163.mm.1 |
MTA TC |