Assay Details
| Gene Symbol: Slc25a23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025877.4 | NP_080153.2 | 4 - 5 | 672 | 62 | |
| GenBank mRNA | AK199713.1 | - | 1 - 2 | 39 | 62 | |
| AK207800.1 | - | 1 - 2 | 49 | 62 | ||
| BC072660.1 | - | 4 - 5 | 668 | 30360302 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a23
Genomic Map