Assay Details
Gene Symbol: Qrich1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114119.1 | NP_001107591.1 | 8 - 9 | 2195 | 69 | |
NM_175143.5 | NP_780352.2 | 7 - 8 | 2071 | 69 | ||
GenBank mRNA | AK151491.1 | - | 8 - 9 | 2169 | 69 | |
AK151530.1 | - | 8 - 9 | 2169 | 69 | ||
AK205435.1 | - | 1 - 2 | 88 | 69 | ||
BC006738.1 | - | 7 - 8 | 2048 | 3156574 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
