| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700023331.mm.1 |
NM_011500.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023331.mm.1 |
NM_011500.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023331.mm.1 |
TC1700002553.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700023333.mm.1 |
NM_011500.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023333.mm.1 |
NM_011500.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023333.mm.1 |
TC1700002553.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700023330.mm.1 |
NM_011500.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023330.mm.1 |
NM_011500.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023330.mm.1 |
TC1700002553.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700023332.mm.1 |
NM_011500.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023332.mm.1 |
NM_011500.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023332.mm.1 |
TC1700002553.mm.1 |
MTA TC |