Assay Details
| Gene Symbol: Strn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011500.2 | NP_035630.2 | 3 - 4 | 480 | 74 | |
| GenBank mRNA | AK051549.1 | - | 3 - 4 | 482 | 74 | |
| BC090968.1 | - | 3 - 4 | 480 | 30646517 | 74 | |
More Information
Set Membership: |
|

Strn
Genomic Map