Assay Details
| Gene Symbol: Kitl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013598.2 | NP_038626.1 | 1 - 2 | 209 | 106 | |
| GenBank mRNA | AK018777.1 | - | 1 - 2 | 209 | 106 | |
| AK134301.1 | - | 1 - 2 | 209 | 106 | ||
| AK165233.1 | - | 1 - 2 | 207 | 106 | ||
| M57647.1 | - | 1 - 2 | 161 | 106 | ||
| M64262.1 | - | 1 - 2 | 81 | 106 | ||
| S40364.1 | - | 1 - 2 | 117 | 106 | ||
| S40534.1 | - | 1 - 2 | 117 | 106 | ||
| S40536.1 | - | 1 - 2 | 117 | 106 | ||
| U44725.1 | - | 1 - 2 | 209 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK018777.1, AK134301.1, AK165233.1, M57647.1, M64262.1, NM_013598.2, S40364.1, S40534.1, S40536.1, U44725.1 transcripts.
Set Membership: |
|

Kitl
Genomic Map