Assay Details
| Gene Symbol: Ppp2r3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161362.3 | NP_001154834.1 | 3 - 4 | 2876 | 151 | |
| NM_172144.3 | NP_742156.2 | 2 - 3 | 716 | 151 | ||
| GenBank mRNA | AK035077.1 | - | 2 - 3 | 496 | 151 | |
| AK076163.1 | - | 2 - 3 | 1928 | 151 | ||
| AK132175.1 | - | 2 - 3 | 717 | 151 | ||
| BC042841.1 | - | 2 - 3 | 1837 | 150 | ||
| BC042842.1 | - | 2 - 3 | 2310 | 150 | ||
| BC151170.1 | - | 2 - 3 | 2703 | 9088059 | 151 | |
| BC151171.1 | - | 2 - 3 | 2698 | 9088060 | 151 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r3a
Genomic Map