Assay Details
| Gene Symbol: Ppp2r3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161362.3 | NP_001154834.1 | 7 - 8 | 3240 | 111 | |
| NM_172144.3 | NP_742156.2 | 6 - 7 | 1080 | 111 | ||
| GenBank mRNA | AK035077.1 | - | 6 - 7 | 860 | 111 | |
| AK076163.1 | - | 6 - 7 | 2292 | 111 | ||
| AK132175.1 | - | 6 - 7 | 1081 | 111 | ||
| BC042841.1 | - | 6 - 7 | 2201 | 111 | ||
| BC042842.1 | - | 6 - 7 | 2674 | 111 | ||
| BC151170.1 | - | 6 - 7 | 3067 | 9088059 | 111 | |
| BC151171.1 | - | 6 - 7 | 3062 | 9088060 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r3a
Genomic Map