Assay Details
Gene Symbol: Tmpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080129.2 | NP_001073598.1 | 7 - 8 | 1290 | 72 | |
NM_001080130.2 | NP_001073599.1 | 6 - 7 | 1170 | 72 | ||
NM_001080131.2 | NP_001073600.1 | 5 - 6 | 1074 | 72 | ||
NM_001283048.1 | NP_001269977.1 | 7 - 8 | 973 | 72 | ||
GenBank mRNA | AA041637.1 | - | 3 - 4 | 301 | 72 | |
AK011988.1 | - | 7 - 8 | 783 | 73 | ||
AK011992.1 | - | 7 - 8 | 783 | 73 | ||
AK141845.1 | - | 7 - 8 | 1240 | 72 | ||
AK146903.1 | - | 7 - 8 | 1257 | 72 | ||
AK150305.1 | - | 7 - 8 | 1238 | 72 | ||
AK150515.1 | - | 7 - 8 | 1225 | 72 | ||
AK151936.1 | - | 7 - 8 | 1242 | 72 | ||
AK152737.1 | - | 7 - 8 | 1242 | 72 | ||
AK153281.1 | - | 7 - 8 | 1225 | 72 | ||
AK159161.1 | - | 7 - 8 | 886 | 72 | ||
AK165282.1 | - | 6 - 7 | 1120 | 72 | ||
AK165851.1 | - | 7 - 8 | 1247 | 72 | ||
AK183465.1 | - | 2 - 3 | 170 | 72 | ||
U39074.1 | - | 7 - 8 | 1221 | 72 | ||
U39075.1 | - | 6 - 7 | 1101 | 72 | ||
U39076.1 | - | 5 - 6 | 1005 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK159161.1, NM_001283048.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
