Assay Details
| Gene Symbol: Adcyap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009625.2 | NP_033755.1 | 1 - 2 | 195 | 63 | |
| GenBank mRNA | BC057344.1 | - | 1 - 2 | 195 | 6829765 | 63 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC057344.1, NM_009625.2 transcripts.
Set Membership: |
|

Adcyap1
Genomic Map