Assay Details
Gene Symbol: Klhdc8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030075.2 | NP_084351.1 | 3 - 4 | 374 | 81 | |
GenBank mRNA | AK019849.1 | - | 3 - 4 | 374 | 81 | |
AK087143.1 | - | 3 - 4 | 360 | 81 | ||
AK154428.1 | - | 3 - 4 | 512 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK019849.1, AK154428.1, NM_030075.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
