Assay Details
Gene Symbol: Klhdc8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030075.2 | NP_084351.1 | 9 - 10 | 2008 | 66 | |
GenBank mRNA | AK019849.1 | - | 9 - 10 | 2006 | 66 | |
AK154428.1 | - | 9 - 10 | 2109 | 66 | ||
BC048918.1 | - | 4 - 5 | 971 | 6309434 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
