Assay Details
Gene Symbol: Klhdc8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030075.2 | NP_084351.1 | 10 - 11 | 2104 | 60 | |
GenBank mRNA | AK019849.1 | - | 10 - 11 | 2102 | 60 | |
AK154428.1 | - | 10 - 11 | 2205 | 60 | ||
BC024793.1 | - | 2 - 3 | 1648 | 60 | ||
BC048918.1 | - | 5 - 6 | 1067 | 6309434 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P609334 | BC048918.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P609334 | BC060089.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P609334 | BC024793.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P609334 | AK154428.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P609334 | NM_030075.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P609334 | BC048918.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P609334 | BC060089.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P609334 | BC024793.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P609334 | AK154428.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P609334 | NM_030075.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 292326 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 820112 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348635 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 351174 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 560396 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 534489 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 90214 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 831508 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 26994 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 102648 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129679 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114180 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P609334 | BC048918.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P609334 | BC060089.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P609334 | BC024793.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P609334 | AK154428.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P609334 | NM_030075.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P609334 | BC048918.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P609334 | BC060089.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P609334 | BC024793.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P609334 | AK154428.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P609334 | NM_030075.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815004 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815019 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814997 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815017 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815001 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815022 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815005 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815013 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814996 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815008 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815023 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815010 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815020 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815015 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815024 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814999 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815000 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814998 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815018 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815011 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26815003 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02026506 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114180 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129679 | - | - |
Whole Mouse Genome G4122A | A_52_P609334 | BC048918.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P609334 | BC060089.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P609334 | BC024793.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P609334 | AK154428.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P609334 | NM_030075.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P609334 | BC048918.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P609334 | BC060089.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P609334 | BC024793.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P609334 | AK154428.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P609334 | NM_030075.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122235 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117715 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114180 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129679 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08088235 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08088230 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08088238 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08088237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814954 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814923 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814909 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814916 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814881 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814957 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814912 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26815013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814964 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814924 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26815010 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07873072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07873075 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814994 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814912 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07908287 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814921 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814926 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814910 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814923 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26815013 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26815011 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814908 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26815003 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814999 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26815024 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814916 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26815000 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26815015 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814905 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814904 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07908292 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814914 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26815022 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08088237 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814918 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26815008 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26815018 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26815026 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814992 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26815001 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26815020 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814903 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08088235 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26815005 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07679230 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26815019 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814993 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10596924 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596917 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596915 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596913 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596920 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596923 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596916 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596922 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596919 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596914 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596918 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596921 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1428221_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10596912 | - | - |
Mouse Genome 430 2.0 Array | 1428221_at | - | - |
Mouse Genome 430 2.0 Array | 1428221_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78267_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78267_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0900025768.mm.1 | NM_030075.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900025768.mm.1 | NM_030075.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900025768.mm.1 | TC0900003061.mm.1 | MTA TC |
Mouse Expression Set 430 | 1428221_PM_at | - | - |
Mouse Expression Set 430 | 1428221_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1428221_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1428221_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 575351 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1216669 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2959360 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2959360 | - | - |
