Assay Details
| Gene Symbol: Parp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK044223.1 | - | 10 - 11 | 1574 | 60 | |
| AK088125.1 | - | 10 - 11 | 1587 | 60 | ||
| BC014870.1 | - | 10 - 11 | 1521 | 3602116 | 60 | |
| BC058754.1 | - | 10 - 11 | 1501 | 6476689 | 60 | |
| BC062204.1 | - | 10 - 11 | 1501 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Parp3
Genomic Map