Assay Details
Gene Symbol: Parp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145619.2 | NP_663594.2 | 7 - 8 | 1161 | 77 | |
GenBank mRNA | AF368233.1 | - | 7 - 8 | 1161 | 77 | |
AK044223.1 | - | 7 - 8 | 1154 | 77 | ||
AK088125.1 | - | 7 - 8 | 1167 | 77 | ||
AK145259.1 | - | 7 - 8 | 1195 | 77 | ||
AK147868.1 | - | 7 - 8 | 1460 | 77 | ||
AK170541.1 | - | 7 - 8 | 1105 | 77 | ||
BC014870.1 | - | 7 - 8 | 1101 | 3602116 | 77 | |
BC058754.1 | - | 7 - 8 | 1081 | 6476689 | 77 | |
BC062204.1 | - | 7 - 8 | 1081 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
