Assay Details
Gene Symbol: Parp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145619.2 | NP_663594.2 | 9 - 10 | 1426 | 111 | |
GenBank mRNA | AF368233.1 | - | 9 - 10 | 1426 | 111 | |
AK044223.1 | - | 9 - 10 | 1419 | 111 | ||
AK088125.1 | - | 9 - 10 | 1432 | 111 | ||
AK145259.1 | - | 9 - 10 | 1460 | 111 | ||
AK147868.1 | - | 9 - 10 | 1725 | 111 | ||
AK165070.1 | - | 7 - 8 | 1676 | 111 | ||
AK170541.1 | - | 9 - 10 | 1370 | 111 | ||
BC014870.1 | - | 9 - 10 | 1366 | 3602116 | 111 | |
BC058754.1 | - | 9 - 10 | 1346 | 6476689 | 111 | |
BC062204.1 | - | 9 - 10 | 1346 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26803652 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26803651 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055487 | AK147868.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055487 | AF368233.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055487 | NM_145619.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 760837 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 427480 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 640367 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 202893 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055487 | AK147868.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055487 | AF368233.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055487 | NM_145619.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803620 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803627 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803614 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803613 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803635 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803648 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803622 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803644 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803633 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803642 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803628 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803624 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803646 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803612 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803636 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803647 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803643 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803615 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803637 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803650 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803631 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803626 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803640 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803651 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803618 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803630 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06915069 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02022960 | - | - |
Whole Mouse Genome G4122A | A_52_P480425 | BC014870.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P480425 | BC058754.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P480425 | AK165070.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P480425 | AK170541.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P480425 | AK145259.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P375069 | AK165070.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P375069 | AK170541.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P375069 | AY046317.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129638 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07853629 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07853622 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26803652 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26803651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803566 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803557 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803564 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803612 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803620 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803599 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803637 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803615 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803651 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803636 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803589 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803626 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803615 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803599 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803637 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07749697 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07911282 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803650 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803640 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08019036 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07638556 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803594 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07638551 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07638553 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07749703 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803635 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803600 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07789198 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803605 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803644 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07817527 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803595 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07817521 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07749695 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07749708 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803628 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803610 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803620 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803618 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803631 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803647 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803646 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803622 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803606 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803648 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803586 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803593 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08146727 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803602 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10596493 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596505 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596501 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596496 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596502 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596494 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596495 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596500 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596497 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596498 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596504 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596503 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596499 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1445888_PM_x_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1426210_PM_x_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451969_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10596492 | - | - |
Mouse Genome 430 2.0 Array | 1451969_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1445888_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1445888_x_at | - | - |
Mouse Genome 430 2.0 Array | 1426210_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1426210_x_at | - | - |
Mouse Genome 430 2.0 Array | 1451969_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 235587_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 235587_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0900025011.mm.1 | NM_145619.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900025011.mm.1 | TC0900003011.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900025015.mm.1 | NM_145619.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900025015.mm.1 | TC0900003011.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900025010.mm.1 | NM_145619.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900025010.mm.1 | TC0900003011.mm.1 | MTA TC |
Mouse Expression Set 430 | 1426210_x_at | - | - |
Mouse Expression Set 430 | 1451969_PM_s_at | - | - |
Mouse Expression Set 430 | 1451969_s_at | - | - |
Mouse Expression Set 430 | 1426210_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1426210_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1426210_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1451969_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1451969_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451969_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451969_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426210_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426210_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1445888_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1445888_PM_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 604623 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1500750 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001500750 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1249773 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1249773 | - | - |
mouse-6 v1.1 (Illumina) | scl35393.12_269-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1500750 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl35393.12_269-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1500750 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2870302 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2870302 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl35393.12_269-S | - | - |
