Assay Details
| Gene Symbol: Slc25a41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175333.3 | NP_780542.1 | 5 - 6 | 667 | 70 | |
| GenBank mRNA | AK077111.1 | - | 5 - 6 | 668 | 70 | |
| BC115588.1 | - | 6 - 7 | 766 | 40102382 | 70 | |
| BC115589.1 | - | 5 - 6 | 660 | 40102384 | 70 | |
| FM165286.1 | - | 5 - 6 | 609 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a41
Genomic Map