Assay Details
| Gene Symbol: Slc25a41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175333.3 | NP_780542.1 | 6 - 7 | 811 | 78 | |
| GenBank mRNA | AK077111.1 | - | 6 - 7 | 812 | 78 | |
| BC115588.1 | - | 7 - 8 | 910 | 40102382 | 78 | |
| BC115589.1 | - | 6 - 7 | 804 | 40102384 | 78 | |
| FM165286.1 | - | 6 - 7 | 753 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331827 | NM_175333.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331827 | AK077111.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331827 | BX637138.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331827 | NM_175333.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331827 | AK077111.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331827 | BX637138.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 598100 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874104 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 460325 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 586638 | AK082669.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 659956 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 128397 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84483 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180411 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331827 | NM_175333.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331827 | AK077111.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331827 | BX637138.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331827 | NM_175333.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331827 | AK077111.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331827 | BX637138.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259709 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259725 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259706 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08813138 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P180411 | - | - |
| Whole Mouse Genome G4122A | A_51_P331827 | NM_175333.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331827 | AK077111.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331827 | BX637138.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331827 | NM_175333.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331827 | AK077111.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331827 | BX637138.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137995 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123960 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P180411 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13408989 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13408990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13308984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259716 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259718 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259712 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13408989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13059709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259742 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13503994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13428393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259734 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259728 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259733 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13164667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259709 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10452254 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10452251 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10452249 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10452256 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10452250 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10452253 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10452255 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10452252 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439756_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10452248 | - | - |
| Mouse Genome 430 2.0 Array | 1439756_at | - | - |
| Mouse Genome 430 2.0 Array | 1439756_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 103775_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 103775_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700021976.mm.1 | NM_175333.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021976.mm.1 | NM_175333.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021976.mm.1 | TC1700002345.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700021977.mm.1 | NM_175333.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021977.mm.1 | NM_175333.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021977.mm.1 | TC1700002345.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439756_PM_at | - | - |
| Mouse Expression Set 430 | 1439756_at | - | - |
| Murine Genome U74v2 Set | 166731_at | - | - |
| Murine Genome U74v2 Set | 166731_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439756_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439756_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 377478 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6860408 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006860408 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1220879 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220879 | - | - |
| mouse-6 v1.1 (Illumina) | scl49751.6.1_171-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6860408 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49751.6.1_171-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6860408 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1220879 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1220879 | - | - |

Slc25a41
Genomic Map