Assay Details
| Gene Symbol: Dpy30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146222.1 | NP_001139694.1 | 1 - 2 | 180 | 163 | |
| GenBank mRNA | BX518196.1 | - | 1 - 2 | 195 | 163 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001146222.1 transcripts.
Set Membership: |
|

Dpy30
Genomic Map