Assay Details
| Gene Symbol: Slc30a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252478.1 | NP_001239407.1 | 9 - 10 | 619 | 58 | |
| NM_144798.6 | NP_659047.2 | 9 - 10 | 604 | 58 | ||
| GenBank mRNA | AF233346.1 | - | 8 - 9 | 556 | 58 | |
| AF395840.1 | - | 8 - 9 | 546 | 58 | ||
| AK082907.1 | - | 9 - 10 | 581 | 58 | ||
| AK150342.1 | - | 9 - 10 | 580 | 58 | ||
| AK151120.1 | - | 9 - 10 | 580 | 58 | ||
| AK160903.1 | - | 9 - 10 | 608 | 58 | ||
| BC005753.1 | - | 1 - 2 | 67 | 58 | ||
| BC066162.1 | - | 9 - 10 | 571 | 30471027 | 58 | |
| BC068169.1 | - | 9 - 10 | 593 | 30542277 | 58 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC005753.1 transcripts.
Set Membership: |
|

Slc30a6
Genomic Map