Assay Details
| Gene Symbol: Gnao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK147783.1 | - | 623 | 118 | ||
| Gene Symbol: Camkmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028576.2 | NP_082852.1 | 6 - 7 | 585 | 118 | |
| GenBank mRNA | AK155246.1 | - | 6 - 7 | 586 | 118 | |
| BC138272.1 | - | 6 - 7 | 733 | 8861291 | 118 | |
| BC138273.1 | - | 6 - 7 | 733 | 8861292 | 118 | |
More Information
Set Membership: |
|

Gnao1
Genomic Map