Assay Details
| Gene Symbol: Mettl4-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016064.1 | - | 566 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016064.1 transcripts.
Set Membership: |
|

Mettl4-ps1
Genomic Map