Assay Details
| Gene Symbol: Galnt14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027864.2 | NP_082140.2 | 3 - 4 | 791 | 101 | |
| GenBank mRNA | AB175681.1 | - | 3 - 4 | 399 | 101 | |
| AK002747.1 | - | 3 - 4 | 763 | 101 | ||
| AK049598.1 | - | 3 - 4 | 737 | 101 | ||
| AK078292.1 | - | 3 - 4 | 404 | 101 | ||
| BC117800.1 | - | 3 - 4 | 575 | 40092267 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt14
Genomic Map