Assay Details
Gene Symbol: Srsf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195485.1 | NP_001182414.1 | 1 - 2 | 126 | 73 | |
NM_001195486.1 | NP_001182415.1 | 1 - 2 | 126 | 73 | ||
NM_001195487.1 | NP_001182416.1 | 1 - 2 | 126 | 73 | ||
NM_146083.2 | NP_666195.1 | 1 - 2 | 126 | 73 | ||
NR_036615.1 | 1 - 2 | 126 | 73 | |||
GenBank mRNA | AK034952.1 | - | 1 - 2 | 128 | 73 | |
AK082848.1 | - | 1 - 2 | 126 | 73 | ||
AK137795.1 | - | 1 - 2 | 127 | 73 | ||
AK161652.1 | - | 1 - 2 | 126 | 73 | ||
AK168355.1 | - | 1 - 2 | 126 | 73 | ||
BC014857.1 | - | 1 - 2 | 107 | 2646366 | 73 | |
BC024865.1 | - | 1 - 2 | 117 | 73 | ||
BC025529.1 | - | 1 - 2 | 117 | 5342587 | 73 | |
BC027391.1 | - | 1 - 2 | 114 | 4945988 | 73 | |
BY740981.1 | - | 1 - 2 | 125 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK082848.1,AK137795.1,AK168355.1,BC014857.1,BC025529.1,BC027391.1,NM_001195485.1,NM_001195486.1,NM_001195487.1,NM_146083.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
