Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700023604.mm.1 |
NM_001195485.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023604.mm.1 |
NM_001195485.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023604.mm.1 |
NM_001195486.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023604.mm.1 |
NM_001195486.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023604.mm.1 |
NM_001195487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023604.mm.1 |
NM_001195487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023604.mm.1 |
NM_146083.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023604.mm.1 |
NM_146083.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023604.mm.1 |
NR_036615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023604.mm.1 |
NR_036615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023604.mm.1 |
TC1700002583.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700023606.mm.1 |
TC1700002583.mm.1 |
MTA TC |