Assay Details
| Gene Symbol: Idh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK190762.1 | - | 84 | |||
| Gene Symbol: Txnrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK146125.1 | - | 10 - 11 | 1225 | 84 | |
| AK168356.1 | - | 11 - 12 | 1332 | 84 | ||
| AK206356.1 | - | 1 - 2 | 48 | 84 | ||
| BC037643.1 | - | 10 - 11 | 1504 | 5059831 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Idh2
Genomic Map