Assay Details
| Gene Symbol: Txnrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042513.1 | NP_001035978.1 | 7 - 8 | 778 | 60 | |
| NM_001042514.1 | NP_001035979.1 | 6 - 7 | 866 | 60 | ||
| NM_001042523.1 | NP_001035988.1 | 7 - 8 | 995 | 60 | ||
| NM_015762.2 | NP_056577.2 | 6 - 7 | 671 | 60 | ||
| GenBank mRNA | AB027565.1 | - | 6 - 7 | 698 | 60 | |
| AF333036.1 | - | 7 - 8 | 995 | 60 | ||
| AK011902.1 | - | 7 - 8 | 779 | 60 | ||
| AK146125.1 | - | 6 - 7 | 672 | 60 | ||
| AK149625.1 | - | 7 - 8 | 779 | 60 | ||
| AK151927.1 | - | 7 - 8 | 779 | 60 | ||
| AK168356.1 | - | 7 - 8 | 779 | 60 | ||
| AK172383.1 | - | 5 - 6 | 1104 | 60 | ||
| AK186228.1 | - | 1 - 2 | 75 | 60 | ||
| BC020274.1 | - | 6 - 7 | 633 | 60 | ||
| BC037643.1 | - | 6 - 7 | 951 | 5059831 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Txnrd1
Genomic Map