Assay Details
| Gene Symbol: Txnrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042513.1 | NP_001035978.1 | 8 - 9 | 1006 | 93 | |
| NM_001042514.1 | NP_001035979.1 | 7 - 8 | 1094 | 93 | ||
| NM_001042523.1 | NP_001035988.1 | 8 - 9 | 1223 | 93 | ||
| NM_015762.2 | NP_056577.2 | 7 - 8 | 899 | 93 | ||
| GenBank mRNA | AB027565.1 | - | 7 - 8 | 926 | 93 | |
| AF333036.1 | - | 8 - 9 | 1223 | 93 | ||
| AK011902.1 | - | 8 - 9 | 1007 | 93 | ||
| AK149625.1 | - | 8 - 9 | 1007 | 93 | ||
| AK151927.1 | - | 8 - 9 | 1007 | 93 | ||
| AK168356.1 | - | 8 - 9 | 1007 | 93 | ||
| AK183038.1 | - | 1 - 2 | 108 | 93 | ||
| AK185436.1 | - | 1 - 2 | 108 | 93 | ||
| AK192872.1 | - | 1 - 2 | 108 | 93 | ||
| AK198860.1 | - | 1 - 2 | 108 | 93 | ||
| AK203979.1 | - | 1 - 2 | 108 | 93 | ||
| AK204938.1 | - | 1 - 2 | 108 | 93 | ||
| BC020274.1 | - | 7 - 8 | 861 | 93 | ||
| BC037643.1 | - | 7 - 8 | 1179 | 5059831 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Txnrd1
Genomic Map