Assay Details
| Gene Symbol: Adgre1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010130.4 | NP_034260.1 | 8 - 9 | 989 | 75 | |
| GenBank mRNA | AK133202.1 | - | 8 - 9 | 965 | 75 | |
| AK149898.1 | - | 8 - 9 | 975 | 75 | ||
| AK151303.1 | - | 8 - 9 | 961 | 75 | ||
| AK151682.1 | - | 8 - 9 | 981 | 75 | ||
| AK155862.1 | - | 8 - 9 | 940 | 75 | ||
| BC075688.1 | - | 8 - 9 | 961 | 30609706 | 75 | |
| U66888.1 | - | 8 - 9 | 976 | 75 | ||
| U66889.1 | - | 8 - 9 | 976 | 75 | ||
| U66890.1 | - | 6 - 7 | 679 | 75 | ||
| U66891.1 | - | 6 - 7 | 700 | 75 | ||
| U66892.1 | - | 5 - 6 | 553 | 75 | ||
| X93328.1 | - | 8 - 9 | 985 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adgre1
Genomic Map