Assay Details
| Gene Symbol: Pja2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025309.1 | NP_001020480.1 | 2 - 3 | 204 | 97 | |
| NM_144859.2 | NP_659108.1 | 2 - 3 | 204 | 97 | ||
| GenBank mRNA | AF493070.1 | - | 2 - 3 | 261 | 97 | |
| AK122282.1 | - | 2 - 3 | 163 | 97 | ||
| AK144586.1 | - | 2 - 3 | 191 | 97 | ||
| AK164025.1 | - | 2 - 3 | 209 | 97 | ||
| AK168371.1 | - | 2 - 3 | 186 | 97 | ||
| BC017130.1 | - | 1 - 2 | 130 | 4506259 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF493070.1, AK144586.1, AK168371.1, BC017130.1, NM_001025309.1, NM_144859.2 transcripts.
Set Membership: |
|

Pja2
Genomic Map