Assay Details
| Gene Symbol: Pja2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025309.1 | NP_001020480.1 | 8 - 9 | 2058 | 86 | |
| NM_144859.2 | NP_659108.1 | 7 - 8 | 1872 | 86 | ||
| GenBank mRNA | AF493070.1 | - | 8 - 9 | 2115 | 86 | |
| AK122282.1 | - | 8 - 9 | 2017 | 86 | ||
| AK144586.1 | - | 8 - 9 | 2045 | 86 | ||
| AK168371.1 | - | 7 - 8 | 1854 | 86 | ||
| BC004742.1 | - | 5 - 6 | 902 | 86 | ||
| BC017130.1 | - | 6 - 7 | 1798 | 4506259 | 86 | |
More Information
Set Membership: |
|

Pja2
Genomic Map