Assay Details
| Gene Symbol: Esco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC008220.1 | - | 5 - 6 | 733 | 131 | |
| BC172153.1 | - | 7 - 8 | 2939 | 131 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Esco1| Gene Symbol: Esco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC008220.1 | - | 5 - 6 | 733 | 131 | |
| BC172153.1 | - | 7 - 8 | 2939 | 131 | ||
Set Membership: |
|