Assay Details
| Gene Symbol: Esco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081222.1 | NP_001074691.1 | 2 - 3 | 428 | 136 | |
| GenBank mRNA | AK020858.1 | - | 2 - 3 | 229 | 136 | |
| AK040576.1 | - | 2 - 3 | 186 | 136 | ||
| AK045656.1 | - | 2 - 3 | 150 | 136 | ||
| AK139549.1 | - | 2 - 3 | 152 | 136 | ||
| AK145333.1 | - | 2 - 3 | 429 | 136 | ||
| BC076613.1 | - | 2 - 3 | 158 | 136 | ||
| BC151069.1 | - | 2 - 3 | 384 | 9087982 | 136 | |
| BC151077.1 | - | 2 - 3 | 384 | 9087990 | 136 | |
| BC172153.1 | - | 2 - 3 | 384 | 136 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK045656.1, AK145333.1, BC151069.1, BC151077.1, NM_001081222.1 transcripts.
Set Membership: |
|

Esco1
Genomic Map