Assay Details
| Gene Symbol: Galm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176963.4 | NP_795937.1 | 3 - 4 | 800 | 92 | |
| GenBank mRNA | AK040998.1 | - | 3 - 4 | 801 | 92 | |
| AK158120.1 | - | 3 - 4 | 599 | 92 | ||
| AK161687.1 | - | 3 - 4 | 599 | 92 | ||
| BC028818.1 | - | 3 - 4 | 603 | 5135570 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galm
Genomic Map