Assay Details
| Gene Symbol: Dhx57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163759.1 | NP_001157231.1 | 6 - 7 | 2014 | 62 | |
| NM_198942.2 | NP_945180.2 | 5 - 6 | 1855 | 62 | ||
| GenBank mRNA | AK053628.1 | - | 6 - 7 | 1682 | 62 | |
| BC062952.1 | - | 5 - 6 | 1855 | 6853031 | 62 | |
| BC065169.1 | - | 6 - 7 | 1681 | 6831314 | 62 | |
| BC066091.1 | - | 5 - 6 | 1517 | 6841207 | 62 | |
| BC079618.1 | - | 5 - 6 | 794 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dhx57
Genomic Map