Assay Details
| Gene Symbol: Magt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190409.1 | NP_001177338.1 | 8 - 9 | 1032 | 64 | |
| NM_025952.4 | NP_080228.4 | 8 - 9 | 1032 | 64 | ||
| GenBank mRNA | AK013243.1 | - | 8 - 9 | 913 | 64 | |
| AK018623.1 | - | 8 - 9 | 935 | 64 | ||
| AK144696.1 | - | 8 - 9 | 911 | 64 | ||
| BC003881.1 | - | 8 - 9 | 919 | 3496572 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Magt1
Genomic Map