Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900009195.mm.1 |
NM_001081041.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900009195.mm.1 |
NM_001081041.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900009195.mm.1 |
TC1900001765.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900009191.mm.1 |
NM_001081041.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900009191.mm.1 |
NM_001081041.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900009191.mm.1 |
TC1900001765.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900009194.mm.1 |
NM_001081041.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900009194.mm.1 |
NM_001081041.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900009194.mm.1 |
TC1900001765.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900009197.mm.1 |
NM_001081041.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900009197.mm.1 |
NM_001081041.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900009197.mm.1 |
TC1900001765.mm.1 |
MTA TC |